Avantor Sterile Polycarbonate Single-Use Bottle Assemblies, Avantor Fluid Handling
Supplier: VWR International
5000005 5000006 5000007 5000008 5000009 5000310
10830-302CS 3034.57 USD
10830-302 10830-304 10830-306 10830-696 10830-698 75835-990
Avantor Sterile Polycarbonate Single-Use Bottle Assemblies, Avantor Fluid Handling
Bottles Cell Culture Bottles
Sterile polycarbonate containers with weldable tubing provide the maximum amount of flexibility and reliability.
  • Sterile bottles come individually bagged and process ready
  • Multiple bottle sizes allow for process-specific flexibility
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
These vessels are assembled in an ISO Class 7 cleanroom and are manufactured from USP Class VI materials.
When used with sterile Avantor tube sets, custom possibilities are endless.
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and cleanroom applications. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to 65 °C (–4 to 149 °F) without losing integrity. All assemblies are individually double-bagged with product label located on inner bag. Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶.
They are offered in a comprehensive size selection with tubing that is completely weldable/sealable.