Avantor Single-Use Tubing Assemblies, Avantor Fluid Handling
Supplier: VWR International
8000002
10830-258CS 1666.72 USD
10830-258
Avantor Single-Use Tubing Assemblies, Avantor Fluid Handling
Tubing Peristaltic Pump Tubing
Unique features make these tubing assemblies a superior alternative to others.
  • Manufactured in ISO Class 7 Cleanroom
  • Gamma irradiated to Sterility Assurance Level (SAL) 10⁻⁶
  • Pinch clamps installed
Assemblies have been designed with a broad spectrum of applications in mind, including life sciences and general lab use, environmental assays, and production and cleanroom applications.
Manufactured from USP Class VI materials. Tubing assemblies are offered with custom size and configuration capabilities. Tubing is weldable/sealable. All product contact materials are animal-free or processed in accordance to EMEA/410/01. The assemblies are able to withstand temperatures from –20 to +65 °C (–4 to +149 °F) without losing integrity. All assemblies are individually double-bagged with product label located on inner bag.
When used with sterile Avantor bottle or flask assemblies, custom possibilities are endless.