Deuterium Lamp for Agilent HPLC Systems, Restek
Supplier: Restek
25888
10830-256EA 1721.99 USD
10830-256
Deuterium Lamp for Agilent HPLC Systems, Restek
Lights Deuterium Lamps
Deuterium lamp, 1100 DAD/MWD/1200, for HPLC systems has life span of 2000 hours.