Gastroccult® Gastric Occult Blood and pH Test System, Beckman Coulter®
Supplier: BECKMAN COULTER
66115A 66040A
10830-226CS 798.68 USD
10830-226 80078-665
Gastroccult® Gastric Occult Blood and pH Test System, Beckman Coulter®
Clinical Diagnostic Supplies Clinical Reagents and Kits
Reliably verifies low levels of blood in samples containing bile.
  • pH comparison chart included
  • Verify blood in bile samples
  • No need for subjective inspection
No need to rely on a subjective visual inspection. A convenient pH comparison chart is included on the test card. The pH scale provides the entire clinically relevant range for monitoring antacid prophylaxis.
Certifications: CLIA waived.
Accessories information: Components are sold separately.