Reveal® G4 Rapid HIV-1/2 Antibody Test, Point-of-Care (POC), MedMira
Supplier: MEDMIRA
Reveal® G4 Rapid HIV-1/2 Antibody Test uses MedMira Rapid Vertical Flow Technology to deliver instant results in three easy steps.
Reveal® G4 POC enables healthcare professionals at point-of-care settings to test easily for HIV, deliver clear results, and begin treatment immediately if needed. The test features a built-in reagent control line, works with fingerstick whole blood specimens, and requires no additional equipment.
HIV-1 sensitivity: 99.64% Fingerstick Whole Blood
HIV-2 sensitivity: 100%.
Specificity: 99.71% Fingerstick Whole Blood
Test kits can be stored for 12 months at 2 to 30°C (35 to 85°F).
Reveal® G4 is a solution for fast and easy testing in POC settings including:
Public health programs
Voluntary testing and counseling centers
Pharmacies and physician offices
Convenience care clinics
Emergency and first responders
HIV-1/2 Antibody Test Controls are designed for use with Reveal® G4 Rapid HIV-1/2 Antibody Test in customer quality assurance practices. Each set of test controls contains 1 HIV-1 Positive, 1 HIV-2 Positive and 1 HIV-1/2 Negative test control. Each vial is sufficient to perform five Reveal® G4 tests. HIV-1/2 Antibody Test Controls have a shelf life of 12 months, and are stored at room temperature until ready for use.
This simple test provides instant results for either reactive or non-reactive specimens 5 times faster than any lateral flow test. Primary applications for Reveal® G4 include occupational exposures, preventing mother-to-child transmission, and emergency departments.
Certifications: FDA Approved.
Ordering information: Each box of Reveal® G4 POC includes 20 individual test kits. Other formats of Reveal® G4 are available for testing venipuncture whole blood, serum, and plasma specimens. Test contols are available as an accessory. Each set of HIV-1/2 Antibody Test Controls contains1 HIV-1 Positive, 1 HIV-2 Positive and 1 HIV-1/2 Negative test control, each sufficient to run five Reveal® G4 tests.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...