PREempt™ Concentrate Disinfectant, Contec®
Supplier: Contec
21305
10822-520CS 224.02 USD
10822-520 10822-522
PREempt™ Concentrate Disinfectant, Contec®
Cleaning Agents Disinfectants
PREempt™ Concentrate Disinfectant Solution works fast for large areas to ensure user, protocol, and product compliance with fast contact times and broad-spectrum efficacy
  • 3 minute Broad-Spectrum Sanitizing (1:128)
  • 30 second Broad-Spectrum Sanitizing (1:16)
PREempt™ Concentrate utilizes AHP®, a patented synergistic blend of commonly used and safe ingredients which, when combined with low levels of hydrogen peroxide, dramatically increase the potency and cleaning performance.
The concentrated formula requires dilution and is cost effective for large surfaces.