DO Electrode, LE621, Mettler-Toledo
Supplier: Mettler Toledo
30266975
10821-390EA 715 USD
10821-390
DO Electrode, LE621, Mettler-Toledo
Electrodes Dissolved Oxygen Electrodes
Galvanic dissolved oxygen sensor with 2 m cable and integrated temperature sensor.
  • IP67 - For a waterproof measurement
  • Integrated temperature probe guarantees you always measure with the appropiate temperature compensation
For use with portable FiveGo meters.