Conductivity Electrode, LE703, Mettler-Toledo
Supplier: Mettler Toledo
30266974
10821-388EA 372 USD
10821-388
Conductivity Electrode, LE703, Mettler-Toledo
Electrodes Conductivity Sensors
Robust conductivity probe for medium to high conductivities with integrated temperature sensor (10 μS/cm–200 mS/cm).
  • IP67 - For a waterproof measurement
  • High accuracy at medium to high conductivities
  • 4-pole cell is a perfect solution to measure a wide range of conductivity samples
  • Integrated temperature probe guarantees you always measure with the appropiate temperature compensation
For use with the portable FiveGo meters.