VWR® Filling and Venting Cap for VWR® Carboys
Supplier: VWR International
583127
10820-438CS 597.29 USD
10820-438
VWR® Filling and Venting Cap for VWR® Carboys
Closures Safety Caps and Safety Waste Caps
The Filling and Venting Cap for VWR® Carboys is an autoclavable closure designed for the aseptic transfer of media, biological reagents, pure water, and chemicals
Each cap is equipped with integrated 1/4" ports.