VWR® Flammable Material Storage Freezers
Supplier: VWR International
10820-434
10820-434EA 2912.14 USD
10820-434
VWR® Flammable Material Storage Freezers
Freezers Flammable Material Freezers
The VWR® Series Flammable Freezers are designed to meet the requirements of NFPA 45 and NFPA 30 for cold storage of flammable and combustible materials in unclassified laboratory locations.
  • Temperature controller
  • Digital setting accuracy
  • Precise temperature maintenance
  • Independent operation for security
  • Monitoring and alarms
  • Meet NFPA guidelines
No electrical components are located in the interior of the unit, and the electrical components of the compressor have sealed in a vapor-proof enclosure for additional safety.
The 113 L (4 cu. ft.) unit uses Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
Units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
Certifications: UL/C-UL listed.
Ordering information: Two-year parts and labor warranty.