VWR® Spatulas, PS
Supplier: VWR International
10820-353
10820-353PK 289.12 USD
10820-353
VWR® Spatulas, PS
Spatulas
Spatula with its long, ergonomic solid handle and pointed blade can be easily pierced into paper and plastic bags.
  • Ideal for the sampling of powder, granules and pastes
  • Individually packed, gamma radiation sterilized
Certifications: Production, assembling and packaging according to cleanroom Class 7 (10,000). Complies with food and FDA guidelines.