Fungi-Fluor® Kit for Fungal Detection, Polysciences
Supplier: POLYSCIENCES INC-RESEARCH CHEM
17442-1
10819-090KT 657.59 USD
10819-090
Fungi-Fluor® Kit for Fungal Detection, Polysciences
Stains Histology Stains
The Fungi-Fluor® Kit for Fungal Detection offers a quick fluorescent stain/counterstain procedure for various fungal organisms
Kit contains 75mL of staining solution A, and 75mL of counterstaining solution B.
The kit can be used to screen a variety of specimen types, such as sputum and skin scrapings, for fungal detection.