Fungi-Fluor® Pneumocystis Kit, Polysciences
Supplier: POLYSCIENCES INC-RESEARCH CHEM
22363-1
10818-624KT 832.18 USD
10818-624
Fungi-Fluor® Pneumocystis Kit, Polysciences
Stains Histology Stains
The Fungi Fluor® Pneumocystis Kit offers a fast, fluorescent staining procedure for Pneumocystis jiroveci in bronchial specimens.
Kit contains 10mL of Fungi-Fluor® staining solution, 10 positive unstained control slides, and Direct fluorescent stain for Pneumocystis carinii cyst.