Plant Isolate DNA Extraction Kit, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J67858-AE
10816-208EA 296.65 USD
10816-208
Plant Isolate DNA Extraction Kit, Thermo Scientific
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Plant isolate kit is CTAB and chloroform based method to isolate total DNA (including genomic, mitochondrial, and chloroplast DNA) from a variety of plant species (including algae and cyanobacteria). The extracted DNA is suitable for routine PCR screening, real-time PCR, southern blotting, mapping, and RFLP.