KLEENGUARD® A20 Breathable Particle Protection Hoods
Supplier: Kimberly-Clark
36890
10815-978CS 273.9 USD
10815-978
KLEENGUARD® A20 Breathable Particle Protection Hoods
Disposable Garments
Made from a patented Microforce™ Barrier SMS Fabric that offers barrier protection against dry particulates and light liquid sprays
The lightweight and breathable properties of the fabric make these hoods comfortable to wear, while the strength and abrasion-resistant properties extend the use of this product. NFPA 99 Compliant Antistatic Material.