1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose 98%
Supplier: Thermo Scientific Chemicals
J66818-03
J66818-06
10815-868EA
37.8
USD
10815-868
10815-870
10815-872
1,2,3-Tri-O-acetyl-5-deoxy-β-D-ribofuranose 98%
1,2,3-Tri-O-acetyl-5-deoxy-beta-D-ribofuranose
Purity: 98%
CAS No.: 62211-93-2
Molecular Formula: C11H16O7
Formula Weight: 260.24
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Melting Point: 65-69°
Solubility: Soluble in ethanol
MDL No.: MFCD08458459
Formula:
C₁₁H₁₆O₇ Melting Pt: 65…69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08458459 CAS Number: 62211-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...