Cycloheximide 90%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,

J66665-06 J66665-03
10814-746EA 195.52 USD
10814-746 10815-830
Cycloheximide 90%
Cycloheximide

A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis.


Purity: 90% CAS No.: 66-81-9 Molecular Formula: C15H23NO4 Formula Weight: 281.35 Flash Point: 110°(230°F) Physical Form: Powder Melting Point: 111-116° Boiling Point: 492° Solubility: Soluble in water at 20mg/ml Beilstein Reference: 88868 Merck Reference: 14,2728 UN No.: 2811 MDL No.: MFCD00082346


A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis.

Formula: C₁₅H₂₃NO₄
Boiling Pt: 492 °C
Melting Pt: 111…116 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082346
CAS Number: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1,III
Merck Index: 14,2728
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR