Lactose
Supplier: Thermo Scientific Chemicals
Synonyms:
Milk sugar, D-Lactose, Anhydrous Lactose, (2R,3R,4R,5R)-2,3,5,6-tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal
J66376-36
J66376-A1
10814-598EA
118.86
USD
10814-598
10814-600
Lactose
Lactose
CAS No.: 63-42-3
Molecular Formula: C12H22O11
Formula Weight: 342.30
Physical Form: Powder
Appearance: White to off-white
Odor: Odorless
Beilstein Reference: 93796
Merck Reference: 14,5343
MDL No.: MFCD00151251
Formula:
C₁₂H₂₂O₁₁ Storage Temperature: Ambient |
MDL Number:
MFCD00151251 CAS Number: 63-42-3 EINECS: 200-559-2 Merck Index: 14,5343 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...