3-Bromo-7-nitro-1H-indazole 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromo-7-nitro-1H-indazole, 3-Bromo-7-nitroindazole

J63480-03 J63480-06
10814-288EA 134 USD
10814-288 10814-290 10814-292
3-Bromo-7-nitro-1H-indazole 98+%
3-Bromo-7-nitro-1H-indazole

Inhibitor of nitric oxide synthase (NOS)


Purity: 98+% CAS No.: 74209-34-0 Molecular Formula: C7H4BrN3O2 Formula Weight: 242.00 Storage Temperature: 2°C to 8°C Physical Form: Powder Sensitivity: Light Sensitive Appearance: Pale yellow Solubility: Soluble in ethanol at 25mg/ml. Also soluble in DMSO MDL No.: MFCD00159910


Inhibitor of nitric oxide synthase (NOS)

Formula: C₇H₄BrN₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00159910
CAS Number: 74209-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR