Hettich MIKRO 185 Microliter Centrifuges, Hettich Lab Technology
Supplier: HETTICH INSTRUMENTS
185MICRO12 185MICRO12-PED 1203-01 185MICROSC 185MICRO24
10813-136EA 3414.48 USD
10813-136 10813-138 75801-794 76300-110 76300-112
Hettich MIKRO 185 Microliter Centrifuges, Hettich Lab Technology
Centrifuges Microcentrifuges
Hettich Lab Technology's MIKRO 185 tabletop microliter centrifuge delivers application versatility and fast results in an elegant, compact frame.
  • Max. Capacity–24 x 1.5/2.0 ml with 24 place rotor rotor
  • Max RPM/RCF–13,300/17,008
  • Ultra-compact
  • All-metal containment
  • Pulse button for quick spins
  • Quiet operation–quiet as a normal conversation, will not increase noise levels or contribute to distractions
  • Functional design–easy to lift lid with front-facing control panel for effortless interaction and programming
  • Enhanced safety–self-closing, auto-locking lid
More Product Information