Spectrophotometer UV5Nano, UV/VIS Excellence, METTLER TOLEDO®
Supplier: Mettler Toledo
30254729
10811-204EA 12111.4 USD
10811-204
Spectrophotometer UV5Nano, UV/VIS Excellence, METTLER TOLEDO®
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The Spectrophotometer UV5Nano is ideal for life science applications such as concentration and purity determinations of DNA, RNA and protein samples using A280, A260, A260/A280, Bradford and BCA protein assays and more.
  • Micro-volume and Kinetics measurements