Contamination Control Swab, Mini Foam Head, Semi-Flexible, Foamtec International WCC
Supplier: Foamtec International
HT1002
10810-864CS 2485.98 USD
10810-864
Contamination Control Swab, Mini Foam Head, Semi-Flexible, Foamtec International WCC
Swabs Swabs for Cleaning
Designed to meet demanding cleanroom applications
Can be used with alcohol, Freon® 113 fluorocarbon, acetone, and other solvents for assembly cleaning, and for epoxy application. Swab features a 100ppi polyurethane head, a semi-flexible tip and a polypropylene handle.
Ordering information: Packaged in polybags.