FirmWIPE® Cleanroom Sponge Wipes, Foamtec International
Supplier: Foamtec International
HT4331
10810-808CS 1502.3 USD
10810-808
FirmWIPE® Cleanroom Sponge Wipes, Foamtec International
Wipes Cleanroom Wipes Dry Cleanroom Wipes
FirmWIPE® is constructed from an open-cell ester-based polyurethane foam which has undergone a special felting process.
  • Open-cell polyurethane structure allows for fiber-free wiping
  • Excellent pick-up of Thick Film inks and solder paste
  • Open-cell, felted foam structure allows for controlled transfer of solvent to optimize cleaning of Thick Film screens
  • Ester-based chemistry and felted construction enable broad solvent compatibility
  • Uniform porosity allows for excellent contamination pickup
FirmWIPE® wipes provides fiber-free wiping in applications where durability and fast wicking, without dripping, are desirable. Due to its ability to conform to metal mask surfaces, wipe enables excess paste to be thoroughly removed while minimizing abrasion to the emulsion. Its unique pore structure makes it ideal for cleaning metal masks used to print LTCC components, capacitors, EL, OLED, and Plasma Display Panels.
Certifications: Cleanroom Class 100 configurations.