Contamination Control Swabs, Foam Head, Firm Tip, Foamtec International WCC
Supplier: Foamtec International
HT1207
10810-776CS 3580.21 USD
10810-776
Contamination Control Swabs, Foam Head, Firm Tip, Foamtec International WCC
Swabs Swabs for Cleaning
Designed to meet demanding cleanroom applications
Can be used with alcohol, Freon® 113 fluorocarbon, acetone and other solvents for assembly cleaning, and for epoxy application. Firm-tip swab features a 100ppi polyurethane head and a wooden handle.
Ordering information: Packaged in polybags.