Brady® Lightweight Universal Absorbent Pads, Brady® SPC® Absorbents
Supplier: Brady Worldwide
107707
10810-754CS 193.72 USD
10810-754
Brady® Lightweight Universal Absorbent Pads, Brady® SPC® Absorbents
Spill Control Materials
General Purpose MAXX® Enhanced absorbents are economical yet offer the look, feel, and performance of a premium product.
  • Highly versatile, grey absorbents pick up water-based, petroleum-based, and non-aggressive chemical fluids
  • Lightweight design specifically targets smaller leaks, drips, and spills
  • General purpose absorbents are made with MAXX® Technology for superior absorbency and greater strength without the added weight
  • Perforations provide cost control and reduce waste by allowing users to utilize only the amount needed
Made with MAXX® for greater performance without the extra weight. Universal in nature, so it absorbs basically all fluids (water, petroleum, and chemical-based). Efficient one-ply construction.
Certifications: EPA 40 CFR 112.7(c)(1)(vii). OSHA 29 CFR 1910.120(j)(1)(vii). OSHA 29 CFR 1910.22(a)(2).