VWR® Air Jacketed CO₂ Incubator, Mini
Supplier: VWR International
10810-742
10810-742EA 6452.5 USD
10810-742
VWR® Air Jacketed CO₂ Incubator, Mini
Incubators CO₂ Incubators
The VWR® Air Jacketed CO2 Incubator Mini is ideal for use as a personal incubator or for low volume applications in a space constrained working environment.
  • 1.4 cu. ft. internal volume
  • Easy-to-clean with coved corners and minimized internal surfaces
  • Supplied with three removable stainless steel shelves and water pan
  • LED display of temperature and CO2 with audible and visual alarm system
  • 3 Year Part and Labor Warranty
Ideal for small-volume applications. Feel safe with a VWR® CO2 incubator that provides you with selection, performance and quality needed in today’s cell culture laboratories. VWR's family of CO2 incubators include water jacketed and air jacketed configurations to meet specific culturing preferences and a wide range of solutions to meet capacity and budget needs. A choice of platform designs and capacities are available to meet your specific culturing preferences. Options include capacities ranging from 1.4 to 6.5 cu. ft., water jacketed models for outstanding temperature stability, and low maintenance air jacketed models with high temperature decontamination cycles. Choice of small capacity models for low volume culturing needs. Stackable models are available to maximize lab space. Choose between thermal conductivity or infrared CO2 sensors to meet preference and application needs.These advanced technologies and contamination control provide a safe growth environment for your cultures. Feel confident that your cultures are in a stable and protected growth environment. Exceptional value for your lab. Fan assisted airflow designed for rapid recovery and uniform conditions. In-chamber probes and sensors designed for precise control. Dual temperatures for over-temperature protection. Polished, high grade stainless steel chambers.