Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® 2™ for infiltration and embedding
Supplier: CANCER DIAGNOSTICS, INC.
PARA2
10810-014CS
487.91
USD
10810-014
Paraffin wax blended with synthetic polymers, Solidification point: 56-58°C, PureAffin® 2™ for infiltration and embedding
Paraffin wax blended with synthetic polymers
PureAffin is specially formulated to provide ideal infiltration with all types of tissue specimens.
Use for both processing and embedding or with PureAffin® 1 Infiltration Medium as part of a two step system. Compatible with all industry paraffins. Blended to provide thorough impregnation of tissues and facilitate excellent sectioning characteristics. Use with open or closed tissue processors. A compound of purified paraffin and additives.
- Stable at elevated temperatures
- Melting Point 56-58°C (133-136F)
- Working Temperature 60°C (140F)
- Water Bath temperature 35-44°C (95-111F)
- Store Below 30°C (86F)
CAS Number:
8002-74-2 |
![]() |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...