Eosin Y (yellowish) solution, Endure™ stain for histology
Supplier: CANCER DIAGNOSTICS, INC.
|
Danger
|
Synonyms:
Solvent Red 43, Field's Stain B, Eosin 225, 2,4,5,7-Tetrabromofluorescein disodium salt, Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoate, Disodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Tetrabromofluorescein disodium salt, 2′,4′,5′,7′-Tetrabromofluorescein disodium salt, Eosin Y disodium salt
The Endure Staining System is designed to produce precise, consistent H&E staining.
Staining times in hematoxylin and eosin will depend on the use and the personal preference of the diagnosing physician. CDI has guidelines that will help establish a custom protocol for your specific needs. The Endure H&E System includes Hematoxylin ED1, (or ED5 “XT” for darker staining), Eosin ED2, Bluing ED3 and Clarifier (Aqueous) ED4 or Clarifier (Alcoholic) ED6. Each component is analytically optimized for staining consistency and longevity.
Formula:
C₂₀H₆Br₄Na₂O₅ MW: 691.86 g/mol |
MDL Number:
MFCD00005040 CAS Number: 17372-87-1 UN: 1170 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...