CDI's Moist Mark Plus Marker, Cancer Diagnostics
Supplier: CANCER DIAGNOSTICS, INC.
MP1000
10809-808PK 161.7 USD
10809-808
CDI's Moist Mark Plus Marker, Cancer Diagnostics
Pens Lab Markers
CDI's Moist Mark Plus Marker writes smooth, dark and fine on all cassettes and all slides.
  • Resistant to all solvents
  • Use for cytology
  • Dotting reference marker
  • Formulated ink
Resistant to all solvents. Writes smooth, dark and fine on all cassettes and all slides. Use for cytology as a "dotting" reference marker as well. Strictest manufacturing standards. Proprietary, formulated ink.