One-Step Blue® Protein Gel Stain, Biotium
Supplier: Biotium
21003-1L
10809-758EA 172.26 USD
10809-758
One-Step Blue® Protein Gel Stain, Biotium
Stains Electrophoresis Stains
One-Step Blue® is a ready-to-use protein gel staining solution. It produces fast (5-60 min) protein staining in a single step without fixation or washing.
  • One-step staining procedure, no fixation required
  • Non-toxic formulation is safe for easy disposal
  • Sensitive protein detection down to 10-20 ng
  • Visualize blue bands, or image with a near-infrared fluorescence imager
  • Replacement for Coomassie stain
  • Store at 4°C
One-Step Blue® is a ready-to-use protein gel staining solution. It produces fast (5-60 min) protein staining in a single step without fixation or washing. Proteins can be detected by visible blue staining, or by near-infrared fluorescence. In addition to rapid results and simple staining, One-Step Blue® offers safer handling and disposal compared to Coomassie staining because it is entirely aqueous-based, without hazardous methanol or acetic acid.
One-Step Blue® has comparable sensitivity as the widely used Coomassie Blue, with a lower limit of detection around 10-20 ng depending on the detection method used. Note that staining intensity varies between proteins. The staining is fully compatible with mass spectrometry and Edman-based sequencing.
Biotium also offers One-Step Lumitein™ Protein Gel Stain, a rapid, easy-to-use, non-toxic red fluorescent protein gel stain for detection using a UV transilluminator or laser gel scanner, that costs significantly less than other fluorescent protein gels stains.