VWR® Premium Webster Needle Holder
Supplier: VWR International
10807-436
10807-436EA 84.3 USD
10807-436
VWR® Premium Webster Needle Holder
Forceps
The VWR® Premium Webster Needle Holder is constructed of surgical-grade German steel.
  • Passivated
  • Fully autoclavable
  • Box-lock joint
  • Rockwell hardness (HRC): 40–45
These reusable forceps can be sterilized by EtO/gamma radiation.