VWR® Luer Bone Rongeur
Supplier: VWR International
11647-232
10806-420EA 345.56 USD
10806-420
VWR® Luer Bone Rongeur
Bone Cutters and Rongeurs
The VWR® Luer Bone Rongeur is made from ASTM (899-94) Series 400 German stainless steel.
  • Passivated
  • Surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
The device is designed for cutting cartilage and bone. A strong, removable bar-spring makes these shears self-opening.