VWR® Petri Dish Sterilization Box
Supplier: VWR International
10806-404
10806-404EA 178.37 USD
10806-404
VWR® Petri Dish Sterilization Box
Trays/Dishes/Pots Laboratory Trays/Dishes
The VWR® Petri Dish Sterilization Box is made from polished ASTM standard (899-94) rugged stainless steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Each box accommodates 18 glass Petri dishes sized 100x10mm (3.93×0.39"), or 10 dishes sized 100x15mm (3.93×0.59").