VWR® Crucible Tongs with PTFE-Coated Tips
Supplier: VWR International
10806-316
10806-316EA 89.56 USD
10806-316
VWR® Crucible Tongs with PTFE-Coated Tips
Tongs
VWR® Crucible Tongs with PTFE Coated Tips are made from ASTM Standard (899-94) Series 400 Stainless Steel.
  • Passivated
  • Long neck serrated tips
  • Raised rivet joint
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
The PTFE-coated tips are designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.