VWR® Jumbo Crucible Tongs
Supplier: VWR International
10806-310
10806-310EA 225.68 USD
10806-310
VWR® Jumbo Crucible Tongs
Tongs
VWR® Jumbo Crucible Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.
  • Passivated
  • Long neck serrated tips
  • Raised rivet joint
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
The Jumbo design facilitates use when reaching into extreme hot or cold environments. These tongs feature integral thumb and finger openings.