VWR® Bone Cutting Shears
Supplier: VWR International
10806-300
10806-300EA 44.77 USD
10806-300
VWR® Bone Cutting Shears
Bone Cutters and Rongeurs
Surgical Grade Stainless Steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods