VWR® Dissecting Bone Cutting Forceps
Supplier: VWR International
10806-298
10806-298EA 31.03 USD
10806-298
VWR® Dissecting Bone Cutting Forceps
Bone Cutters and Rongeurs
Surgical Grade Stainless Steel.
  • Passivated
  • Compatible with Gamma/ETO/Autoclavable sterilization methods