VWR® Beyer Double-Action Bone Rongeur
Supplier: VWR International
10806-262
10806-262EA 342.22 USD
10806-262
VWR® Beyer Double-Action Bone Rongeur
Bone Cutters and Rongeurs
The VWR® Beyer Double-Action Bone Rongeur is made from ASTM (899-94) Series 400 German stainless steel
  • Passivated
  • Surgical grade
  • Rockwell Hardness (HRC): 50–55
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
The instrument is designed for cutting cartilage and bone, and features a strong, removable bar-spring that makes the shears self-opening.