VWR® Dish Tongs
Supplier: VWR International
11647-160
10806-214EA 45.01 USD
10806-214
VWR® Dish Tongs
Tongs
VWR® Dish Tongs are made from ASTM Standard (899-94) Series 400 Stainless Steel.
  • Passivated
  • Rounded jaws
  • Raised riveted joint
  • Compatible with Gamma/ETO/Autoclavable sterilization methods
Tongs are designed to safely and easily handle evaporating dishes, hollow articles, and more. Tongs feature rounded jaws that provide a 7.6 to 12 cm (3 to 4.75") diameter opening to accommodate vessels from 70 mL to 250 mL.