PurSwab® Anti-Static Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1806-PFAS
10806-154CS 956.97 USD
10806-154
PurSwab® Anti-Static Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Cleaning
These swabs feature a completely static-free construction
This swab is used as a general cleaning tool during electronic assembly and medical device manufacturing, and for removing contamination in static sensitive components. It is ideal for applying adhesives or other solvents, is lint- and residue-free, and can be used dry or with IPA and DI water. Made in USA.
They incorporate an antistatic polypropylene shaft and foam head. Static-dissipative swabs attract dirt and dust for easy cleaning, while reducing and dissipating static electricity. Antistatic properties will not decay over time.
Certifications: CE marked.
Ordering information: Swabs are packaged 50 swabs per bag, 20 bags per case.