Puritan® Glue Free Cotton Tipped Applicator, Wooden Handle, Puritan Medical Products
Supplier: Puritan Medical Products
867-WCNOGLUE 838-WCS-NG
10806-005CS 532.69 USD
10806-005 10805-132
Puritan® Glue Free Cotton Tipped Applicator, Wooden Handle, Puritan Medical Products
Swabs Swabs for Sampling
Swab features a special construction that eliminates chemical migration.
Wood shaft. Adhesive-free.
Certifications: CE marked.
Ordering information: Packaged 100 swabs per bag, 10 bags per box, 10 boxes per case.