UltraPlex 1-Step ToughMix (4X), QuantaBio
Supplier: Quantabio
UltraPlex 1-Step ToughMix is a ready-to-use, 4X concentrated master mix 1-step reverse transcription and real-time quantitative PCR (RT-qPCR) of RNA templates using probe-based detection methods. First-strand cDNA synthesis and PCR amplification are carried out in the same tube without opening between steps. Optimized to deliver highly sensitive quantification of RNA viruses or low abundance RNA targets in uni- or highly multiplexed RNA detection assays, this reagent chemistry is optimized to deliver maximum assay sensitivity, precision and reproducibility with miniaturized reaction volumes and either conventional or accelerated thermal cycling protocols. UltraPlex 1-Step ToughMix contains all required components for RT-qPCR except RNA template and probe and is compatible with all dual-labeled probe chemistries.
- 4X concentrated master mix allows greater flexibility with diluted samples (as little as 1 pg of total RNA) and multiplexing up to 5 targets
- Tough-tested for applications where PCR inhibition from template impurities may be a concern
- Temperature stabilized master mix enables convenient reaction setup at room temperature
UltraPlex 1-Step ToughMix (Low ROX/ ROX) (4X), contains:
4X reaction buffer containing optimized titrations of qPCR-grade dATP, dCTP, dGTP, dTTP, and magnesium.
Ultra-pure recombinant proteins: qScript XLT reverse transcriptase, RNase inhibitor protein, and AccuStart II hot-start Taq DNA polymerase.
Proprietary stabilizers additives and optimized ROX passive reference dye (if applicable).
The system has been optimized to deliver maximum RT‑qPCR efficiency, sensitivity, and specificity in reduced reaction volumes and fast cycle times. UltraPlex 1-Step ToughMix contains all required components for RT-qPCR except RNA template and probe. It is compatible with all dual-labeled probe chemistries.
Caution: UltraPlex 1-Step ToughMix is intended for molecular biology applications. This product is not intended for the diagnosis, prevention or treatment of a disease. Store components in a constant temperature freezer at -25°C to -15°C protected from light upon receipt.