Medifuge™ Small Benchtop Centrifuge, Thermo Scientific
Supplier: Thermo Fisher Scientific
75008801
10803-926EA 2777.75 USD
10803-926
Medifuge™ Small Benchtop Centrifuge, Thermo Scientific
Centrifuges Benchtop Centrifuges
Perform both routine clinical and life science separations with the Medifuge™ Small Benchtop Centrifuge
  • Promote safety and environmental sustainability in the lab:
  • Emergency lid lock release available in case of power failure
  • Quiet operation at noise levels less than 56 dBA
  • Lightweight composite rotor is easy to handle and saves energy during acceleration/deceleration
  • Conforms to the latest clinical and safety standards, such as UL, CE, and IVD
The lightweight 8-place rotor features a unique hybrid design with interchangeable fixed angle and swinging buckets, and is able to accommodate 1.4 mL to 15 mL tubes with two spacer options. Run common clinical IVD blood tubes, gel tubes, 10 mL standard syringes and 15 mL conical tubes—all in one versatile centrifuge with just one rotor.
It fits in with its compact design and stands out with the unique DualSpin™ rotor, featuring a 2-in-1 hybrid design. Easily change between fixed angle and swinging buckets using the same rotor. Four customizable programs allow users to quickly run routine protocols. Three selectable deceleration profiles, including brake-off option, are designed to optimize separation.
Certifications: UL listed, CE marked, RoHS, CSA, IVD, FDA class 1 compliant.
IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101, EN 61326-1, EN ISO 13485
Ordering information: Each Medifuge™ Centrifuge packages includes the Medifuge™ Small Benchtop Centrifuge, DualSpin™ rotor body, fixed angle buckets (set of 8), swinging buckets (set of 8), green/short spacers (set of 8), and yellow/long spacers (set of 8).