Easy Na Sodium Analyzer
Supplier: Mettler Toledo
30060051
10803-512EA 12140 USD
10803-512
Easy Na Sodium Analyzer
Analyzers Elemental Analysers
The Easy Na Sodium analyzer from METTLER TOLEDO determines the sodium content in food and other products simply and quickly.
  • Easy to use
  • Smartphone-style apps design
  • One LongClick™ needed
  • Save money
  • No calibration necessary
  • Simple result data management
Reduce sample preparation time and effort by using safer and less expensive chemicals while, specifically and exclusively measuring sodium ions.
Replace the silver nitrate (AgNO₃) titrant normally used for chloride determination with the less expensive and safer sodium reagents. Due to the standard addition technique procedure, no calibration is necessary. Just carry out the automated sensor conditioning and immediately start the analysis. Connect your sodium analyzer to EasyDirect™ Titration Software or to a USB-P25 dot-matrix printer.
Simply add a sample and ionic strength adjustment (ISA) solution, and start the measurement. Smartphone-style apps design is used to controls all major functions. One LongClick™ is all that is necessary to start the last analysis run.
Certifications: UL listed and CSA certified. IEC and EN certified.
Ordering information: Standard delivery–sodium analyzer with analyzer head, EasyStir and magnetic stir bar, 10 ml burette, bottle head for sodium standard, DX222-Na Sodium ISE, DX205-SC reference electrode and cable, tubing set, 110 V power adapter, EasyDirect data transfer software (trial), and Application Brochure.