Mouse Follistatin
Supplier: Rockland Immunochemical
010-001-U88-0100 010-001-U88-0020 010-001-U88-0005
10801-450EA 8070 USD
10801-450 10801-454 10802-938 10802-942
Mouse Follistatin
Proteins and Peptides
Recombinant Mouse Follistatin control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Follistatin is an autocrine acting protein that is expressed by many tissues, but at noteably higher levels in the ovary and skin. Follistatin functions to negatively regulate the signaling of a wide variety of TGF-beta family members (activin, BMPs, myostatin, GDF-11 and TGF-beta 1). Mechanistically, follistatin works as an antagonist by complexing with TGF-beta family members to prevent them from interacting with their signaling receptors. Recombinant mouse follistatin is a non-glycosylated protein, containing 289 amino acids, with a molecular weight of 31.6 kDa.
Follistatin Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Follistatin in immunological assays.