Human Recombinant Flt-3 Ligand
Supplier: Rockland Immunochemical
009-001-U86-0010 009-001-U86-0100
10802-912EA 6397 USD
10802-912 10802-916 10802-920 10802-924
Human Recombinant Flt-3 Ligand
Proteins and Peptides
Recombinant Human Flt-3 Ligand control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
FMS-related Tyrosine Kinase 3 Ligand (FLT-3 Ligand) is a growth factor important for the proliferation of hematopoietic cells. FLT-3 Ligand binds to, and transmits signals through, the receptor tyrosine kinase known as FMS-like Tyrosine Kinase-3 (FLT-3). FLT-3 Ligand promotes long-term expansion and differentiation of human pro-B cells in the presence of IL-7 or in combination of IL-7 and IL-3. Human FLT-3 Ligand can stimulate the proliferation of cells expressing murine FLT-3 receptors. Recombinant human FLT-3 Ligand is a non-glycosylated protein, containing 155 amino acids, with a molecular weight of 17.6 kDa.
Flt-3 Ligand Recombinant Protein is suitable as a control for polyclonal or monoclonal anti- Flt-3 Ligand in immunological assays. Buffer Formulation: 10 mM sodium phosphate and 50 mM sodium chloride, pH 7.5.