Anti-VPS51 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-BD7
10802-752EA 635.22 USD
10802-752
Anti-VPS51 Rabbit Polyclonal Antibody
Antibodies
Fat Free, also known as ANG2, was identified though a yeast two-hybrid screen using VP53. It is thought to be a structural and functional homolog of the yeast protein VPS51, a protein that together with VPS52, VPS53, and VPS54 form the Golgi-associated retrograde protein (GARP) complex that mediates the tethering and fusion of endosome-derived transport carriers to the trans-Golgi network (TGN). Similar to depletion of any of the VPS52, VPS53, or VPS54, depletion of Fat Free using RNAi impairs protein retrieval to the TGN indicating that Fat Free is missing component of the GARP complex in most eukaryotes.

Recommended Dilutions: ELISA: 1:10,000; Western Blot: 1 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: VPS51
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR