Human Recombinant FADD (from E. coli)
Supplier: Rockland Immunochemical
009-001-R92S
10802-740EA 380.3 USD
10802-740 10802-742
Human Recombinant FADD (from E. coli)
Proteins and Peptides
FADD or Fas-Associated protein with Death Domain is an adaptor molecule that mediates death signaling by the Fas-receptor, tumor necrosis factor receptor and TRAIL-receptor. FADD binds to these receptors via the C-terminus Death Domain which then unmasks the N-terminal effector domain of FADD thereby allowing it to recruit caspase-8 and activate the cysteine protease cascade leading to apoptosis (1). Cells lacking FADD are defective in intracellular double-stranded RNA (dsRNA)-activated gene expression, including production of type I (alpha/beta) interferons and are thus very susceptible to viral infection (2). FADD Protein is ideal for investigators involved in Signaling Proteins, Adaptor Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, and NfkB Pathway research.