Anti-IgG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
314-4102
10802-730EA 311.34 USD
10802-730
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR