Anti-ENDOG Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-BA5
10802-622EA 635.22 USD
10802-622
Anti-ENDOG Rabbit Polyclonal Antibody
Antibodies
The fragmentation of nuclear DNA is a hallmark of apoptotic cell death. The activities of caspase and nuclease are involved in the DNA fragmentation. Caspase-activated deoxyribonuclease (CAD), also termed DNA fragmentation factor (DFF40), is one such nuclease, and is capable of inducing DNA fragmentation and chromatin condensation after cleavage by caspase-3 of its inhibitor ICAD/DFF45. Caspase and CAD independent DNA fragmentation also exists. Recent studies demonstrated that another nuclease, endonuclease G (endoG), is specifically activated by apoptotic stimuli and is able to induce nucleosomal fragmentation of DNA independently of caspase and DFF/CAD. EndoG is a mitochondrion-specific nuclease that translocates to the nucleus and cleaves chromatin DNA during apoptosis. The homologue of mammalian EndoG is the first mitochondrial protein identified to be involved in apoptosis in C. elegans. EndooG also cleaves DNA in vitro.

Recommended Dilutions: ELISA: 1:20,000; Immunohistochemsitry: 15 ?g/mL; Western Blot: 2 ?g/mL; contains 0.02% (w/v) Sodium Azide

Type: Primary
Antigen: ENDOG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR